(4-Chlorphenyl)acetone 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Chlorophenylacetone

350700050 350700250
ACRO350700050EA 791 DKK
ACRO350700050 ACRO350700250
(4-Chlorphenyl)acetone 98%
(4-Chlorphenyl)acetone
Formel: C₉H₉ClO
Kogepunkt: 132 °C (12 mmHg)
Massefylde: 1,151 g/cm³
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00045214
CAS-nummer: 5586-88-9
EINECS: 226-986-4

Bestil nu

Specifikationer Testresultater

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5325 to 1.5345 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR