3-Amino-2-methylbenzoesyre 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B24947.06 B24947.14
B24947.06EA 396 DKK
B24947.06 B24947.14
3-Amino-2-methylbenzoesyre 98%
3-Amino-2-methylbenzoesyre

Purity: 98% CAS No.: 52130-17-3 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 185-188° Beilstein Reference: 2803090

Formel: C₈H₉NO₂
Kogepunkt: 347 °C (1013 hPa)
Smeltepunkt: 185…188 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075026
CAS-nummer: 52130-17-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR