Propiophenon for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Phenyl ethyl ketone, Ethyl phenyl ketone, 1-Phenyl-1-propanone
M-Clarity™ quality level = MQ200
8.07024.0250
8.07024.1000
8.07024.0250EA
280
DKK
8.07024.0250
8.07024.1000
Propiophenon for synthesis, Sigma-Aldrich®
Propiophenon
Formel:
C₆H₅C(O)CH₂CH₃ Molekylvægt: 134.18 g/mol Kogepunkt: 217 Smeltepunkt: 18 Massefylde: 1.01 g/cm³ (20 °C) Flammepunkt: 85 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009309 CAS-nummer: 93-55-0 EINECS: 202-257-6 Merck Index: 12,08015 |
|
Specifikationer Testresultater
Solubility | 2 g/l (20 °C) |
Melting Point | 18 °C |
Molar Mass | 134.17 g/mol |
Boiling Point | 214 - 218 °C (1013 hPa) |
Vapor Pressure | 1.99 hPa (20 °C) |
Flash Point | 85 °C |
Refractive Index | 1.5258 (20 °C, 589 nm) |
Density | 1.01 g/cm3 (20 °C) |
Synonyms | 1-Phenyl-1-propanone, Ethyl phenyl ketone |
Hill Formula | C9H10O |
Chemical Formula | C6H5COC2H5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...