Heptanophenon ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A10491.09 A10491.18
A10491.09EA 610 DKK
A10491.09 A10491.18
Heptanophenon ≥98%
Heptanophenon

Purity: 98+% CAS No.: 1671-75-6 Molecular Formula: C13H18O Formula Weight: 190.29 Flash Point: 110°(230°F) Refractive Index: 1.5077 Melting Point: ca 17° Boiling Point: 283° Beilstein Reference: 1865692 MDL No.: MFCD00009539

Formel: C₆H₅C(O)(CH₂)₅CH₃
Kogepunkt: 283 °C
Smeltepunkt: ∼17 °C
Massefylde: 0,946
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009539
CAS-nummer: 1671-75-6
EINECS: 216-802-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fe3edbdffde888a03942372840c87d884038a92&vid=2305382a25957397ce425d97191b024a89fb99bf&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR