Perifer IV-kanyle, BD Angiocath™
Leverandør: BD Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
382277 382259 381164 382258 382268 382268UK 381112 381123 381134 381137 381144 381147 381154 381157 382287
BDAM382277EA 1150 DKK
BDAM382277 BDAM382259 BDAM381164 BDAM382258 BDAM382268 BDAM382268UK BDAM381112 BDAM381123 BDAM381134 BDAM381137 BDAM381144 BDAM381147 BDAM381154 BDAM381157 BDAM382287
Perifer IV-kanyle, BD Angiocath™
Kanyler
The BD Angiocath™ and BD Angiocath™ special IV catheters offer a wide range of products designed to provide peripheral vascular access to help meet clinicians' needs. The catheters are made with FEP material with a clear flash chamber to help make blood flashback easily identifiable.
  • Nål med tredobbelt affasning, latexfri
  • Krystalklart flashback-kammer
  • Luer-slip- og luer-lock-kompatibel