4-Ethylbenzaldehyd ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: p-Ethylbenzaldehyde

B20645.14 B20645.22
B20645.14EA 478 DKK
B20645.14 B20645.22
4-Ethylbenzaldehyd ≥97%
4-Ethylbenzaldehyde

Purity: 97% CAS No.: 4748-78-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 92°(197°F) Refractive Index: 1.5390 Odour: Characteristic Boiling Point: 220-221° Beilstein Reference: 636027 MDL No.: MFCD00006956

Formel: C₉H₁₀O
Kogepunkt: 220…221 °C
Massefylde: 0,979
Flammepunkt: 92 °C (197 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006956
CAS-nummer: 4748-78-1
EINECS: 225-268-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR