2-Methylbenzaldehyd ≥98% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: o-Tolualdehyde, 2-Methylbenzaldehyde

Stabilization: Stabilised with hydroquinone (HQ)

A10172.14 A10172.22
A10172.14EA 533 DKK
A10172.14 A10172.22
2-Methylbenzaldehyd ≥98% stabilised
2-Tolualdehyde

Stabilized with 0.1% hydroquinone


Purity: 98% CAS No.: 529-20-4 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 67°(152°F) Refractive Index: 1.5470 Odour: Almond-like Melting Point: -35° Boiling Point: 199-200° Beilstein Reference: 605841 Merck Reference: 14,9527 MDL No.: MFCD00003338

Formel: C₈H₈O
Molekylvægt: 120.15 g/mol
Kogepunkt: 199…200 °C
Smeltepunkt: –35 °C
Massefylde: 1.037
Flammepunkt: 67 °C (152 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003338
CAS-nummer: 529-20-4
EINECS: 208-452-2
Merck Index: 14,9527

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR