Support clamps
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17210 17211 17212 17213 17214 17215 17216
BOCH17210EA 9830 DKK
BOCH17210 BOCH17211 BOCH17212 BOCH17213 BOCH17214 BOCH17215 BOCH17216
Support clamps
Klemmer Universalklemmer
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
  • For reactors and other vessels
  • User friendly thanks to joint screws
  • With two supporting devices for 26,9 mm tubes