Tromlepumper, gastætte
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5611-1090 5611-0090
223-0071EA 18870 DKK
223-0071 223-0072
Tromlepumper, gastætte
Pumper Tromlepumper
Gas tight barrel pump with flexible discharge hose for filling off in to large canisters and containers.
  • Convenient transfer as the canister can stay on the floor
  • Pump can be hermetically sealed with two ball valves
Leveringsinformation: Includes gas-tight barrel screw joints with 2" fine thread.