α-Brom-2-Fluortoulen ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene
A12692.09
A12692.18
A12692.09EA
502
DKK
A12692.09
A12692.18
α-Brom-2-Fluortoulen ≥98%
α-Brom-2-Fluortoulen
Purity: 98%
CAS No.: 446-48-0
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 82°(179°F)
Refractive Index: 1.5520
Odour: Pungent
Boiling Point: 84-85°/15mm
Beilstein Reference: 1099894
UN No.: 3265
MDL No.: MFCD00000324
Formel:
C₇H₆BrF Kogepunkt: 84…85 °C (15 mmHg) Massefylde: 1,567 Flammepunkt: 82 °C (179 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000324 CAS-nummer: 446-48-0 EINECS: 207-169-1 UN nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...