HPLC columns, Accucore™ RP-MS
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17626-012105 17626-152130 17626-013005 17626-014005 17626-032130 17626-052130 17626-053030 17626-054630 17626-102130 17626-103030 17626-104630 17626-153030 17626-154630
554-1945EA 4570 DKK
554-1945 554-1888 554-1946 554-1947 554-1870 554-1882 554-1883 554-1884 554-1885 554-1886 554-1887 554-1889 554-1890
HPLC columns, Accucore™ RP-MS
Kromatografikolonner
Optimise LC/MS analyses with Thermo Scientific™ Accucore RP-MS columns.
  • Lower back pressures
  • Excellent peak shapes
These columns are optimised for MS detection with an excellent combination of speed and quality of separation. RP-MS offers slightly lower retention than C18 which, when combined with high efficiencies and low peak tailing, makes this the phase of choice for MS detection. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub 2 µm particles.
Bestillingsoplysninger: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder, 2,1/3 mm int.Ø require the 2/3 mm UNIGUARD holder and 4/4,6 mm int.Ø require the 4/4,6 mm UNIGUARD holder.