Stirrers, propeller, with two paddles
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9704 9705 9703
BOCH9704EA 270 DKK
BOCH9704 BOCH9705 441-1908
Stirrers, propeller, with two paddles
Omrørere Omrøreudstyr
Propeller stirrer made of stainless steel with two fixed paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow