Prøvetagningsøser
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5389-0250 5389-0100 5389-1000 5389-0500 5389-0010
BURK5389-0250EA 3510 DKK
BURK5389-0250 BURK5389-0100 BURK5389-1000 BURK5389-0500 BURK5389-0010
Prøvetagningsøser
Prøvetagere Væskeprøvetagere
All parts coming into contact with the sample medium are made of pure white PTFE.
  • Completely inert against all substances (with the exception of elementary fluoride)
  • The rod is stabilised with a steel core which is completely encapsulated in PTFE
  • The 60 cm long rod can be extended to 120 cm by adding another component
  • The scoop PTFE with 10 ml has a strong rod with a length of 155 mm