Klemmer til koniske standardslib, KECK™, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286337302 286337405 286337508 286337602
271-7130EA 80.1 DKK
271-7130 271-7138 271-7135 271-7136
Klemmer til koniske standardslib, KECK™, DURAN®
Klemmer Klemmer til NS-slib
Nickel with steel spring. KECK™ conical joint clips ensure a secure connection and thus increase laboratory safety.
  • Easy to fit and do not alter their shape
  • Ideal for applications with very high temperatures