Serum collection tubes, vacuum, BD Vacutainer®
Leverandør: BD Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
367624 369032 368815 367614 367896 368493 368975 367837 367895UK 367814
613-3974EA 1210 DKK
613-3974 BDAM369032 BDAM368815 BDAM367614 BDAM367896 BDAM368493 BDAM368975 BDAM367837 BDAM367895UK BDAM367814
Serum collection tubes, vacuum, BD Vacutainer®
Kliniske diagnose systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, transparent.
  • With or without additive
  • With label
In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer® Plus plastic serum tubes have silica particles added for this purpose. These tubes are marked with the acronym CAT (Clot Activator Tube).