Bowls
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8787 8786 8789 8780 8781 8782 8783 8784 8785
BOCH8787EA 764 DKK
BOCH8787 BOCH8786 BOCH8789 BOCH8780 BOCH8781 BOCH8782 BOCH8783 BOCH8784 BOCH8785
Bowls
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Made of stainless steel with round bottom.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose bowls ideal for a wide variety of uses.