Propper, diamantslib, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286117407 286118103 286117904 286113808 286114204 286114804 286115406 286116008 286116608
217-0793EA 3100 DKK
217-0793 217-0795 217-0794 217-0787 217-0788 217-0789 217-0790 217-0791 217-0792
Propper, diamantslib, DURAN®
Propper Glas
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Pointed
  • Diamond-ground