Test tubes with screw caps
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
84001 84002 84004 84005 84003 84000 84017 84018 8401600
KART84001EA 715 DKK
KART84001 KART84002 KART84004 KART84005 KART84003 KART84000 KART84017 KART84018 KART8401600
Test tubes with screw caps
Rør Reagensglas
PP, graduated, conical. Tight sealing screw cap made from PP.
  • Withstand up to −80 °C
  • Can be centrifuged up to 3500 ×g (self-standing bottom) and 5000 ×g (without self-standing bottom)
  • Sterile tubes are supplied with a red cap