Slange, gummi, vakuum
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3885 3886 3887 3888
KART3885EA 4840 DKK
KART3885 KART3886 KART3887 KART3888
Slange, gummi, vakuum
Slanger Overførselsslange
Made of rubber, grey.
Hoses with cloth impression surface are suitable for air, water, medium concentration acids and alkaline solutions, abrasive products, oxygenised solvents.
Temperature range: −40 to +70 °C.