4-(Benzyloxy)benzoesyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A15715.14 A15715.03 A15715.06
A15715.14EA 3090 DKK
A15715.14 A15715.03 A15715.06
4-(Benzyloxy)benzoesyre ≥98%
4-(Benzyloxy)benzoesyre

Purity: 98% CAS No.: 1486-51-7 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 187-191° Beilstein Reference: 2111640

Formel: C₁₄H₁₂O₃
Molekylvægt: 228.25 g/mol
Smeltepunkt: 187…191 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016527
CAS-nummer: 1486-51-7
EINECS: 216-066-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR