Methylindol-5-carboxylat ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 1H-indole-5-carboxylate
L16228.06
L16228.03
L16228.06EA
1750
DKK
L16228.06
L16228.03
Methylindol-5-carboxylat ≥98%
Methylindol-5-carboxylat
Purity: 98+%
CAS No.: 1011-65-0
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 123-126°
Beilstein Reference: 474256
Formel:
C₁₀H₉NO₂ Molekylvægt: 175.19 g/mol Smeltepunkt: 123…126 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00153023 CAS-nummer: 1011-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...