Ethyl indol-2-carboxylat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

L00797.06 L00797.14
L00797.06EA 613 DKK
L00797.06 L00797.14
Ethyl indol-2-carboxylat ≥98%
Ethyl indol-2-carboxylat

Purity: 98+% CAS No.: 3770-50-1 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 122-125° Beilstein Reference: 146395

Formel: C₁₁H₁₁NO₂
Smeltepunkt: 122…125 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005609
CAS-nummer: 3770-50-1
EINECS: 223-206-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR