Geraniol

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-ol

4988.1 4988.2
ROTH4988.1EA 0 DKK
ROTH4988.1 ROTH4988.2
Geraniol
Geraniol
Formel: C₁₀H₁₈O
Molekylvægt: 154.25 g/mol
MDL Number: MFCD00002917
CAS-nummer: 106-24-1
EINECS: 203-377-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR