o-Dianisidin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,3'-Dimethoxybenzidin

407890050 407890250
ACRO407890050EA 313 DKK
ACRO407890050 ACRO407890250
o-Dianisidin 97%
o-Dianisidin
Formel: C₁₄H₁₆N₂O₂
Smeltepunkt: 135…139 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008372
CAS-nummer: 119-90-4
EINECS: 204-355-4
ADR: 6.1,II

Bestil nu

Specifikationer Testresultater

Appearance Beige to pink to light brown to purple Crystalline powder or crystals (May darken on storage)
Infrared spectrum Conforms
Melting point 134 to 140 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR