Mycophenolic acid, Millipore®

Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

475913-100 475913-500
475913-100EA 1830 DKK
475913-100 475913-500
Mycophenolic acid, Millipore®
Mycophenolic acid
Formel: C₁₇H₂₀O₆
Molekylvægt: 320.34 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00036814
CAS-nummer: 24280-93-1
EINECS: 246-119-3

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR