Diethylfluormalonat ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 685-88-1
Molecular Formula: FCH(CO2CH2CH3)2
Formula Weight: 178.16
Flash Point: 62°(143°F)
Refractive Index: 1.4070
Boiling Point: 121-122°/30mm
Beilstein Reference: 1775686
UN No.: 1760
MDL No.: MFCD00009139
Formel:
C₇H₁₁FO₄ Kogepunkt: 121…122 °C (30 mmHg) Massefylde: 1,129 Flammepunkt: 62 °C (143 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009139 CAS-nummer: 685-88-1 EINECS: 211-684-7 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...