2-Amino-4-tert-butylphenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A0957-25G
A0957-250G
TCIAA0957-25GEA
288
DKK
TCIAA0957-25G
TCIAA0957-250G
2-Amino-4-tert-butylphenol ≥98.0% (by GC, titration analysis)
2-Amino-4-tert-butylphenol
Formel:
C₁₀H₁₅NO |
MDL Number:
MFCD00007698 CAS-nummer: 1199-46-8 EINECS: 214-844-4 |
|
Specifikationer Testresultater
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 158.0 to 164.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...