DL-Threonin 99.5%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
138940250
138941000
138945000
ACRO138940250EA
376
DKK
ACRO138940250
ACRO138941000
ACRO138945000
DL-Threonin 99.5%
DL-Threonin
Formel:
C₄H₉NO₃ Molekylvægt: 119.12 g/mol Smeltepunkt: 244 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00063722 CAS-nummer: 80-68-2 EINECS: 201-300-6 |
Specifikationer Testresultater
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.2 % (1 g, 105°C, 3 hrs) |
Sulfated ash | ≤0.1 % |
Titration with HClO4 | 98.0 to 101.0 % (On dried substance) |
Chloride (Cl) | ≤200 ppm |
Arsenic (As) | ≤2 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Other amino acids | (TLC) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...