3,5-Dimethoxyphenol ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 500-99-2
Molecular Formula: C8H10O3
Formula Weight: 154.16
Flash Point: 78°(173°F)
Melting Point: 45-47°
Boiling Point: 172-175°/17mm
Beilstein Reference: 1239156
MDL No.: MFCD00008388
Formel:
C₈H₁₀O₃ Kogepunkt: 172…175 °C (17 mmHg) Smeltepunkt: 45…47 °C Flammepunkt: 78 °C (173 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008388 CAS-nummer: 500-99-2 EINECS: 207-917-7 |