Tridekansyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
n-Tridecanoic acid, Tridecylic acid
T0412-25G
T0412-500G
TCIAT0412-25GEA
542
DKK
TCIAT0412-25G
TCIAT0412-500G
Tridekansyre ≥98.0% (by GC, titration analysis)
Tridekansyre
Formel:
C₁₃H₂₆O₂ Kogepunkt: 308 °C (1013 hPa) Massefylde: 1,025 g/cm³ (20 °C) Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00002741 CAS-nummer: 638-53-9 EINECS: 211-341-1 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 40.0 to 43.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...