2-Undecanon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
U0006-25ML
U0006-500ML
TCIAU0006-25MLEA
132
DKK
TCIAU0006-25ML
TCIAU0006-500ML
2-Undecanon ≥98.0% (by GC)
2-Undecanone
Formel:
C₁₁H₂₂O Kogepunkt: 120°C Smeltepunkt: ∼12 °C Flammepunkt: 106°C |
MDL Number:
MFCD00009583 CAS-nummer: 112-12-9 EINECS: 203-937-5 ADR: 9,III Merck Index: 12,06178 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...