Pyrocatechol dimethylether for synthesis

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether

M-Clarity™ quality level = MQ200

8.08529.0250 8.08529.1000
8.08529.0250EA 418 DKK
8.08529.0250 8.08529.1000
Pyrocatechol dimethylether for synthesis
Pyrocatechol dimethylether
Formel: C₈H₁₀O₂
Kogepunkt: 207 °C (1013 hPa)
Smeltepunkt: 19…21 °C
Massefylde: 1,085 g/cm³ (20 °C)
Flammepunkt: 87 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008357
CAS-nummer: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) insoluble
Melting Point 17.5 °C
Molar Mass 138.16 g/mol
Boiling Point 205 - 206 °C
Vapor Pressure 0.63 hPa (25 °C)
Flash Point 87 °C
Refractive Index 1.5346 (20 °C, 589 nm)
Density 1.085 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR