3-Methoxyphenol ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A12826.14 A12826.22 A12826.36
A12826.14EA 341 DKK
A12826.14 A12826.22 A12826.36
3-Methoxyphenol ≥97%

Purity: 97% CAS No.: 150-19-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 124°(255°F) Refractive Index: 1.5520 Odour: Sweet Melting Point: -18° Boiling Point: 244° Beilstein Reference: 1209898 UN No.: 2810 MDL No.: MFCD00002267

Formel: C₇H₈O₂
Kogepunkt: 244 °C
Smeltepunkt: –18 °C
Massefylde: 1,140
Flammepunkt: 124 °C (255 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002267
CAS-nummer: 150-19-6
EINECS: 205-754-6
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR