Komponenter til reaktionsbeholdere
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
210343405 210344607 210345106 210345706 210346102 210353406 210354608 210355107 210355707 210356103
SCOT210343405EA 848 DKK
SCOT210343405 SCOT210344607 SCOT210345106 SCOT210345706 SCOT210346102 SCOT210353406 SCOT210354608 SCOT210355107 SCOT210355707 SCOT210356103
Komponenter til reaktionsbeholdere
Fittings og stik
These flanges are available with or without groove.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Made from clear DURAN® borosilicate glass 3.3.