Water testing reagents and test kits for WTW spectrophotometers and photometers
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
250425 250426 250427 250428 250433 250435 250438 250441 250442 250443 250444 250445 250446 250447 250451 250331 250353 250452 250491 250492 252012 251400 251401 251402 251403 251404 251405 251406 251408 251409 251410 251411 251412 251988 251989 251990 251991 251992 251993 251994 251995 251996 251998 251413 250493 250440 252017 252035 252077 252079 252078 251414 252058 252043 252044 252037 252046 252011 252027 252048 252063 252019 252041 252016 252050 250301 252049 252047 252042 252034 252022 252020 252014 252010 250439 250422 250419 250306 252021 252089 252087 252053 252084 252082 252068 252085 252097 252096
WTWA250425EA 2180 DKK
WTWA250425 WTWA250426 WTWA250427 713-4050 WTWA250433 713-2260 WTWA250438 WTWA250441 WTWA250442 WTWA250443 WTWA250444 WTWA250445 713-2271 WTWA250447 WTWA250451 713-4057 713-4014 713-2277 713-4051 713-4055 WTWA252012 WTWA251400 WTWA251401 WTWA251402 WTWA251403 WTWA251404 WTWA251405 WTWA251406 WTWA251408 WTWA251409 WTWA251410 WTWA251411 WTWA251412 WTWA251988 WTWA251989 WTWA251990 WTWA251991 WTWA251992 WTWA251993 WTWA251994 WTWA251995 WTWA251996 WTWA251998 WTWA251413 WTWA250493 WTWA250440 WTWA252017 WTWA252035 WTWA252077 WTWA252079 WTWA252078 WTWA251414 WTWA252058 WTWA252043 WTWA252044 WTWA252037 WTWA252046 WTWA252011 WTWA252027 WTWA252048 WTWA252063 WTWA252019 WTWA252041 WTWA252016 WTWA252050 WTWA250301 WTWA252049 WTWA252047 WTWA252042 WTWA252034 WTWA252022 WTWA252020 WTWA252014 WTWA252010 WTWA250439 WTWA250422 WTWA250419 WTWA250306 WTWA252021 705-0726 WTWA252087 WTWA252053 WTWA252084 WTWA252082 WTWA252068 WTWA252085 WTWA252097 WTWA252096
Water testing reagents and test kits for WTW spectrophotometers and photometers
Miljø test kit
A wide choice of tests is available, for routine quantitative analysis in different applications, designed for use with WTW spectrophotometers and photometers. The test is automatically recognised by the laboratory instruments (via a barcoding system) and results are displayed in the appropriate units. Due to the different photometer optics, one test may yield different measuring ranges in different instruments; LED photometers may have smaller measuring ranges for the same test.