Pipette leak testing unit
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
703970 703980
613-1557EA 15740 DKK
613-1557 613-5740
Pipette leak testing unit
Pipetter Automatiske pipetter
The PLT (Pipette Leak Tester) unit for air displacement pipettes allows daily pipette checks, providing a safeguard during periods between routine calibrations thereby significantly improving the pipette process reliability. Even the smallest leaks are detected and quantified.
  • Test limit values for all commercially available single- and multi-channel pipettes in the volume range of 1 μl to 10 ml are pre-programmed
  • Test pipettes with and without tips to assist in diagnosis of the leak source
  • Test results in seconds
  • Optional PLT unit software (PLTconnect) facilitates the documentation of leak tests
Bestillingsoplysninger: Supplied with 1 of each type of adapter (4-channel pipette adapter optional) for testing air displacement pipettes with tip (attached) or testing without tip, 2 plugs (for instrument validation), 3 replacement PE filters for the pipette adapters, universal AC adapter, quality certificate and operating manual.