Transfer pipettes, The Dropette®
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P200-58 P200-6020S P200-72 P200-10 P200-52 P200-3020S
SIMPP200-58EA 4030 DKK
SIMPP200-58 SIMPP200-6020S SIMPP200-72 SIMPP200-10 SIMPP200-52 SIMPP200-3020S
Transfer pipettes, The Dropette®
Pipetter Pasteur-/Overførselspipetter
LDPE, disposable. These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
  • Molded from see-through low density polyethylene
  • Inert to biological fluids and most acids
  • No bulb to insert or remove
  • Can be sealed and refrigerated
  • Temperature resistant down to –196 °C
The low-affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
*Graduated.