Laboratorierengøringskoncentrater, rea-sol
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9720101 9720105 9720130
HIRS9720101EA 343 DKK
HIRS9720101 HIRS9720105 HIRS9720130
Laboratorierengøringskoncentrater, rea-sol
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Surface cleaner in a liquid concentrate format that rapidly cleans materials such as glass, porcelain, rubber, plastics and metals of oil, grease, wax, resin, silicone, blood, protein and serum residues. Also suitable for cleaning and decontamination of walls and floors. Environmentally friendly as the active agents are biologically degradable.
  • Graduations on glassware are unaffected
  • Free of chlorine and caustic alkalis
  • Good skin toleration and non toxic
  • Extremely economical: Typically 1,0 % working solution
  • Suitable for ultrasonic cleaning baths
Advarsel: Long exposure times should be avoided for metals such as aluminium and zinc.