N-Ethylanilin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

118040025 118041000 118045000
ACRO118040025EA 1870 DKK
ACRO118040025 ACRO118041000 ACRO118045000
N-Ethylanilin 98%
N-Ethylanilin
Formel: C₈H₁₁N
Kogepunkt: 204,7 °C (1013 hPa)
Smeltepunkt: –63 °C
Massefylde: 0,963 g/cm³
Flammepunkt: 85 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009025
CAS-nummer: 103-69-5
EINECS: 203-135-5
UN nummer: 2272
ADR: 6.1,III
Merck Index: 13,03800

Bestil nu

Specifikationer Testresultater

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5530 to 1.5560 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR