3-Nitroanilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: m-Nitroanilin

A10629.0E A10629.22 A10629.36
A10629.0EEA 3790 DKK
A10629.0E A10629.22 A10629.36
3-Nitroanilin ≥98%
3-Nitroanilin

Purity: 98% CAS No.: 99-09-2 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: 112-114° Boiling Point: 306° Merck Reference: 14,6582 UN No.: 1661 MDL No.: MFCD00007782

Formel: C₆H₆N₂O₂
Molekylvægt: 138.13 g/mol
Kogepunkt: 306 °C
Smeltepunkt: 112…114 °C
Massefylde: 0.901
Flammepunkt: 196
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007782
CAS-nummer: 99-09-2
EINECS: 202-729-1
UN nummer: 1661
ADR: 6.1 II
Merck Index: 12,06679

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR