Test tubes with screw cap
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5620 5600 0560100 5622
KART5620EA 1840 DKK
KART5620 KART5600 KART0560100 KART5622
Test tubes with screw cap
Rør Reagensglas
PS, clear, with PE screw cap.
  • Good thermal resistance
  • Good chemical resistance
Tubes have a round bottom and a freestanding skirt. Sterile tubes are available individually wrapped. The 30 ml individual tubes have a self-adhesive label for marking.