Prøvetagere, engangsbrug, stempelhævert
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5387-0100 5387-0075 5387-0050
BURK5387-0100EA 544 DKK
BURK5387-0100 BURK5387-0075 BURK5387-0050
Prøvetagere, engangsbrug, stempelhævert
Prøvetagere Prøvetagere til engangsbrug
Disposable plunging samplers acting like siphons.
  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler
If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.