N,N-Dimethyl-4-nitroanilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L00404.06 L00404.14
L00404.06EA 389 DKK
L00404.06 L00404.14
N,N-Dimethyl-4-nitroanilin ≥98%
N,N-Dimethyl-4-nitroanilin

Purity: 98+% CAS No.: 100-23-2 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 163-166° Beilstein Reference: 638087 UN No.: 2811 MDL No.: MFCD00014712

Formel: C₈H₁₀N₂O₂
Molekylvægt: 166.18 g/mol
Kogepunkt: 288
Smeltepunkt: 163…166 °C
Massefylde: 1.179 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014712
CAS-nummer: 100-23-2
EINECS: 202-832-1
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR