Emballageflasker, bred åbning, sorte, med skruelåg, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
209683 209685 209686
WHEA209683EA 3460 DKK
WHEA209683 WHEA209685 WHEA209686
Emballageflasker, bred åbning, sorte, med skruelåg, WHEATON®
Flasker Plastflasker
HDPE, translucent.
  • Good chemical resistance
  • Blake design maximizes storage space
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner
Wide mouth ideal for solids.