Cryostorage boxes, KeepIT-100™, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
W651703-B W651703-G W651703-P W651703-R W651703-W W651703-Y W651704-B W651704-G W651704-P W651704-R W651704-W W651704-Y W651702-B W651702-G W651702-P W651702-R W651702-W W651702-Y
479-2104EA 2520 DKK
479-2104 479-2103 479-2101 479-2100 479-2099 479-2102 479-2110 479-2109 479-2107 479-2106 479-2105 479-2108 479-2098 479-2097 479-2095 479-2094 479-2093 479-2096
Cryostorage boxes, KeepIT-100™, WHEATON®
Bokse Æsker til Kryorør
Plastic, coloured.
  • Box inventory eliminates unnecessary entry or removal of sample for scanning
  • Traceability by lots of 100
  • Eliminates handling of individual samples, therefore does not compromise sample integrity
Unique design, featuring punch-holes in bottom of the base to facilicate reading/scanning of 2D bar-coded bottom plugs of CryoELITE™ vial; in addition, quicker draining of Liquid Nitrogen.
The KeepIT-100™ has a transparent notched cover for easy see through sample indentification; solvent resistant grid (1 to 100) withstands cryogenic temperatures and accommodates labels and freezer pens. Clear lid pairs well with coloured capped cryogenic vials; this enable colour coding sample identification without removing lid. Accommodates up to 100 (2 ml) internal thread cryogenic vials. Standard footprint, compatible with liquid nitrogen storage shelves and freezer drawers.