Burettes, straight stopcock, PYREX®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3295/02 3295/04 3295/06 3295/08
612-3955EA 450 DKK
612-3955 612-3956 612-3957 612-3958
Burettes, straight stopcock, PYREX®
Buretter
Borosilicate glass with interchangeable PTFE key, Class B.
  • Calibrated to deliver ‘Ex’
  • PTFE key for grease free performance
  • White enamel graduations for good visibility with dark coloured solutions
Certificeringer: ISO 385