Support rods, polished
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
240.204 240.205 240.206 240.207 240.208 240.209 240.210 241.205 241.210 241.209 241.208 241.206 241.204 241.207
RSGA240.204EA 210 DKK
RSGA240.204 241-0236 RSGA240.206 241-0237 241-0238 RSGA240.209 RSGA240.210 RSGA241.205 RSGA241.210 RSGA241.209 RSGA241.208 RSGA241.206 RSGA241.204 RSGA241.207
Support rods, polished
Stativstænger
Polished rods made from stainless steel 18/10. Available with or without thread.