N,N-Diethylanilin ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A10560.0D
A10560.0F
A10560.AP
A10560.0DEA
4550
DKK
A10560.0D
A10560.0F
A10560.AP
N,N-Diethylanilin ≥99%
N,N-Diethylanilin
Purity: 99%
CAS No.: 91-66-7
Molecular Formula: C10H15N
Formula Weight: 149.24
Flash Point: 85°(185°F)
Refractive Index: 1.5420
Melting Point: -38°
Boiling Point: 215-216°
Beilstein Reference: 742483
Merck Reference: 14,3114
UN No.: 2432
MDL No.: MFCD00009042
Formel:
C₁₀H₁₅N Molekylvægt: 149.24 g/mol Kogepunkt: 215…216 °C Smeltepunkt: –38 °C Massefylde: 0.936 Flammepunkt: 85 °C (185 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009042 CAS-nummer: 91-66-7 EINECS: 202-088-8 UN nummer: 2432 ADR: 6.1 III Merck Index: 13,03140 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...