1-iod-2-nitrobenzen ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A12100.14 A12100.22
A12100.14EA 1020 DKK
A12100.14 A12100.22
1-iod-2-nitrobenzen ≥97%
1-iod-2-nitrobenzen

Purity: 97% CAS No.: 609-73-4 Molecular Formula: C6H4INO2 Formula Weight: 249.01 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 288-289° Beilstein Reference: 2044110 MDL No.: MFCD00007088

Formel: C₆H₄INO₂
Kogepunkt: 288…289 °C
Smeltepunkt: 48…52 °C
Massefylde: 1,9186 g/cm³ (75 °C)
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007088
CAS-nummer: 609-73-4
EINECS: 210-200-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR