3-Iodanilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L01159.09 L01159.18
L01159.09EA 747 DKK
L01159.09 L01159.18
3-Iodanilin ≥98%
3-Iodanilin

Purity: 98% CAS No.: 626-01-7 Molecular Formula: C6H6IN Formula Weight: 219.03 Flash Point: 110°(230°F) Refractive Index: 1.6840 Melting Point: 21-24° Boiling Point: 145-146°/15mm Beilstein Reference: 636058 MDL No.: MFCD00007781

Formel: C₆H₆IN
Molekylvægt: 219.02 g/mol
Kogepunkt: 145…146 °C (15 mmHg)
Smeltepunkt: 21…24 °C
Massefylde: 1.821
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007781
CAS-nummer: 626-01-7
EINECS: 210-922-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR